Learn More
Bioassay Systems QuantiChrom™ Indican Assay Kit. For quantitative determination of indican (indoxyl sulfate) in urine samples.
SDP
Supplier:  Bioassay Systems DIDC100
Encompass_Preferred
For quantitative determination of indican (indoxyl sulfate) in urine samples. Key Features: Fast and sensitive. Linear detection range: 0.2 - 20 mg/dL (8-800 µmoles/L). Fast and high-throughput. Homogeneous "mix-incubate-measure" type assay. Assay takes only 10 min. Can be readily automated for processing thousands of samples per day. Method: OD480nm. Samples: Urine. Species: All. Procedure: Assay takes 10 min. Kit size: 100 tests. Detection limit: 0.2 mg/dL.
Catalog No. 50-489-306