Learn More
QIAGEN QIAcard FTA Indicating Classic
Description
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Classic with 4 sample areas per card.
- QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
- Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
- Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Specifications
| Description | Collect, stabilize, process, transport, and archive buccal DNA samples, four samples per card. |
| Format | Card |
| Sample Type | Buccal DNA |
| For Use With (Equipment) | Direct STR amplification |
| For Use With (Application) | Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology |
| Includes | Pink indicating dye |
| No. of Samples | 4 |
| Product Type | Indicating FTA Classic Card |
| Quantity | 100 Pk. |
| Sample Volume | 125 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.