Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indicator A, Reagents

Catalog No. 01336171
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-171 1 L Glass Bottle
01-336-172 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-171 Supplier Reagents Holdings Llc Supplier No. C84030NE1C
Add to Cart
Add to Cart

Indicator A, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Indicator A
Boiling Point 100°C
CAS 7732-18-5, 3625-57-8
Synonym Nile blue sulfate
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.001
Color Blue
Quantity 1 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.