Learn More
MEDPLUS SERVICES USA INDICATOR INCUBATOR 28-VIAL
Supplier:  MEDPLUS SERVICES USA MMM 127
Encompass_Preferred
• 3M Attest Incubators provide optimal conditions for accurate readout of 3M Attest Biological Indicators 3M Attest Biological Indicator Incubator, 28-vial (2-tier), 37°C EO, 120 nominal voltage required, 90-132V acceptable 3M Attest Biological Indicator Incubator, 28-vial (2-tier), 37°C EO, 120 nominal voltage required, 90-132V acceptable. Attest incubators provide optimal incubation conditions for the accurate readout of 3M™ Attest™ Biological Indicators. Since the growth of each test organism is temperature dependent, it is critical that the appropriate Attest biological indicator be used with the correct Attest incubator.
Catalog No. NC1234719
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.