Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Indicator Solution, Universal
SCI_ED

Catalog No. S25619 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
100 mL
500 mL
Ea.
Packaging:
Color Chart
Solution
Solution with Color Chart
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25619 Ea. Color Chart
S25363B 100 mL Solution
S25363 500 mL Solution
S25363A 1 L Solution with Color Chart
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S25619
pH indicator; for pH determinations from pH 4.0 to 10.0

This product is for educational use - no C of A is available.

Specifications

Chemical Name or Material Universal Indicator Solution
CAS 34722-90-2, 64-17-5, 547-58-0, 845-10-3, 518-51-4
Assay Percent Range <0.2% w/v
Synonym Universal pH Indicator
Packaging Color Chart
Density 0.93
Color Green
Quantity Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.