Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indigo Carmine (Certified Biological Stain), Fisher Chemical

Catalog No. I16625 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I166-25 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. I166-25

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 860-22-0
Molecular Formula C16H8N2Na2O8S2
Molecular Weight (g/mol) 466.35
MDL Number MFCD00005723
InChI Key KHLVKKOJDHCJMG-QDBORUFSSA-L
Synonym Acid Blue 74, FD&C Blue No. 2
PubChem CID 5284351
IUPAC Name disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O

Specifications

Bio. Stain Comm. Certification Pass test
Dye Content 80 % min.
Packaging Glass Bottle
Color Index No. 73015
Color Blue
Quantity 25 g
Physical Form Powder
Formula Weight 466.37
Grade Certified Biological Stain
Chemical Name or Material Indigo Carmine
Safety handling Check Alert storage code Red
Hazard Category Signal Word: WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed.
NFPA
Health: 1
Flammability: 0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.