CAS RN 860-22-0
Thermo Scientific Chemicals Indigo Carmine, pure, certified
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.35 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L Synonym: Acid Blue 74,C.I. 73015,5, 5'-Indigodisulfonic acid, disodium salt PubChem CID: 5284351 IUPAC Name: disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
Indigo Carmine 95.0+%, TCI America™
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.35 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L Synonym: indigo carmine,indigocarmine,acid blue 74,indigocarmin,indigotindisulfonate sodium,c.i. acid blue 74,food blue no. 2,amacid brilliant blue,c.i. food blue 1,fd&c blue no. 2 PubChem CID: 5284351 IUPAC Name: disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
Thermo Scientific Chemicals Indigo carmine
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.35 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L Synonym: indigo carmine,indigocarmine,acid blue 74,indigocarmin,indigotindisulfonate sodium,c.i. acid blue 74,food blue no. 2,amacid brilliant blue,c.i. food blue 1,fd&c blue no. 2 PubChem CID: 5284351 SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
5,5'-Indigodisulfonic Acid, Disodium Salt, BAKER™, J.T. Baker™
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.35 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L Synonym: indigo carmine,indigocarmine,acid blue 74,indigocarmin,indigotindisulfonate sodium,c.i. acid blue 74,food blue no. 2,amacid brilliant blue,c.i. food blue 1,fd&c blue no. 2 PubChem CID: 5284351 IUPAC Name: disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
Indigo Carmine, Reagent, ACS, Spectrum™ Chemical
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.35 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L IUPAC Name: disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
FD and C Blue No. 2, Spectrum™ Chemical
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.35 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L IUPAC Name: disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
Indigo Carmine, MP Biomedicals™
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.35 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L Synonym: Acid Blue 74,Indigo-5, 5'-disulfonic acid disodium salt PubChem CID: 5284351 IUPAC Name: disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
Thermo Scientific Chemicals Indigo Carmine, for analysis, high purity biological stain
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.35 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L Synonym: Acid Blue 74,C.I. 73015,5, 5'-Indigodisulfonic acid, disodium salt PubChem CID: 5284351 IUPAC Name: disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
Food Blue No. 2, TCI America™
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.35 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L PubChem CID: 5284351 IUPAC Name: disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
Indigo Carmine (Acid Blue 74), Reagents
Indigo Carmine (Acid Blue 74), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Fisher Science Education™ Indigo Carmine
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.35 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L Synonym: Carmine Blue (Biological Stain),C.I. 73015 PubChem CID: 5284351 IUPAC Name: disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
Indigo Carmine (Certified Biological Stain), Fisher Chemical
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.35 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L Synonym: Acid Blue 74,FD&C Blue No. 2 PubChem CID: 5284351 IUPAC Name: disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
MP Biomedicals, Inc Indigocarmine, Reag, MP Biomedicals
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.35 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L PubChem CID: 5284351 IUPAC Name: disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O