Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indigo Carmine, MP Biomedicals™

Catalog No. ICN15263825 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15263825 25 g
ICN15263805 5 g
02-152-63880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15263825 Supplier MP Biomedicals, Inc Supplier No. 0215263825
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indigo Carmine is used as a stain.

  • Indigo Carmine is useful in the staining of Negri bodies.
  • It can be used as a contrast stain for plasma when red nuclear stains are used.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 860-22-0
Molecular Formula C16H8N2Na2O8S2
Molecular Weight (g/mol) 466.35
MDL Number MFCD00005723
InChI Key KHLVKKOJDHCJMG-QDBORUFSSA-L
Synonym Acid Blue 74, Indigo-5, 5'-disulfonic acid disodium salt
PubChem CID 5284351
IUPAC Name disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O

Specifications

Absorbance λ max (water) 608 to 610 nm
Assay Percent Range ˜85%
Color Purple
Solubility Information 10 to 50 mg/mL at 70.7°F
Quantity 25 g
Physical Form Powder
Formula Weight 466.3
Grade Certified Biological Stain
Chemical Name or Material Indigo carmine, Cell Biology and Analysis, Molecular Biology
EINECSNumber 212-728-8
storageNote1 Protect from light.
Recommended Storage Store at room temperature.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.