Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Indigo Carmine, pH Indicator and Tissue Stain, 10 g

Catalog No. 502704691
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4691 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4691 Supplier Abcam Supplier No. AB14635210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Versatile pH indicator, tissue stain, and analytical tool for hypochlorite determination.

  • Molecular Weight: 466.4 g/mol
  • Applications: pH indicator (blue at pH 11.4 and yellow at 13.0), tissue stain, and tool for determination of hypochlorite
  • Absorption: λmax 608 nm
  • Form: Solid
  • Composition: 100% Indigo carmine

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Hypochlorite
Shipping Condition Ambient
Content And Storage Store at ambient temperature.
Form Solid
Detection Method Colorimetric
Quantity 10 g
For Use With (Application) pH indicator, tissue staining, hypochlorite determination
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.