Learn More
Abcam Indigo Carmine, pH Indicator and Tissue Stain, 50 g
Description
- Molecular weight: 466.4 g/mol
- Form: Solid
- Applications: pH indicator (blue at pH 11.4 and yellow at 13.0), tissue stain, and tool for the determination of hypochlorite
- Absorption: λmax 608 nm
- Constituents: 100% Indigo carmine
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Hypochlorite |
| Shipping Condition | Ambient |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Detection Method | Colorimetric |
| Formulation | 100% Indigo carmine |
| Quantity | 50 g |
| For Use With (Application) | pH indicator, Tissue stain, Determination of hypochlorite |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.