Learn More
PIP™ Indirect Vent Goggle
Catalog No. 19148213 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Catalog No. Lens Coating
19-148-213 Anti-fog, Scratch-resistant
1 options
This item is not returnable. View return policy
Catalog No. 19-148-213 Supplier PIP™ Supplier No. 251800020RHB
  • $492.50
    $248.65 / Pack of 10
    Save $243.85
    50% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,713.00
    $994.65 / Case of 4 PK
    Save $718.35
    42% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
PIP™ Indirect Vent Goggle offers indirect ventilation for liquid splash protection
  • Wide facial flange for a secure seal
  • Flexible, soft rubber/polyester body for optimal comfort and fit
  • Fits comfortably over most prescription glasses
  • Nose area split to fit the vast majority of users
  • Wide adjustable neoprene headband strap
  • Headband strap attaches to frame with a swivel clip to ensure a comfortable fit
  • FogLess™ lens coating
  • Comes individually boxed
  • 5.25 Base curve lens
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance Meets Z87+ Requirements
Lens Tint Clear
Product Type Indirect Vent Safety Goggles
Frame Color Light Gray
Includes Neoprene strap
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Rubber
Frame Type Indirect Vented
Show More Show Less
Warning - Prop 65 - Reproductive Harm