Learn More
Medchemexpress LLC Indirubin-3′-oxime | 667463-82-3 | 99.1% | C16H11N3O2 | 10MG

Supplier: Medchemexpress LLC HY13925410MG
Indirubin-3′-oxime is a synthetic indirubin derivative used as a research reagent that potently inhibits cyclin-dependent kinases (CDKs), GSK3β, and JNK. Supplied as a brown-red solid with reported purity of 99.1%, it is intended for biochemical kinase assays, cell-based studies, and in vivo formulations; soluble in DMSO and available in multiple milligram sizes. CAS 667463-82-3.
- Potent kinase inhibitor of CDKs, GSK3β, and JNK.
- High purity (99.1%) suitable for research applications.
- Available as solid powder and ready-to-use DMSO solutions.
- Soluble in DMSO at high concentration (approximately 100 mg/mL).
- Storage conditions support long-term stability at -20°C for powder.
- Provided with datasheet, certificate of analysis, and safety data sheet for quality and handling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.