Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) iodide, anhydrous, 99.999% (metals basis)

Catalog No. AA1185409
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1185409 10 g
AA1185404 2 g
AA1185418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1185409 Supplier Thermo Scientific Chemicals Supplier No. 01185409
Add to Cart

CAS: 13510-35-5 | I3In | 495.531 g/mol

Indium(III) iodide is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13510-35-5
Molecular Formula I3In
Molecular Weight (g/mol) 495.531
MDL Number MFCD00016151
InChI Key RMUKCGUDVKEQPL-UHFFFAOYSA-K
Synonym indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
PubChem CID 83539
IUPAC Name triiodoindigane
SMILES [In](I)(I)I

Specifications

Melting Point 210°C
Color Red to Yellow
Physical Form Crystalline
Quantity 10 g
Assay Percent Range 99.994% (Metals basis)
Solubility Information Soluble in water.
Formula Weight 495.53
Sensitivity Hygroscopic
Density 4.69
Chemical Name or Material Indium(III) iodide
Hazard Category H315-H317-H319-H334-H335-H360
Precautionary Statement P201-P202-P261-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P501c
EINECSNumber 236-839-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.