Learn More
Abcam Indo-1 AM, Fluorescent Ca²± Indicator, Cell-permeable, 1 mg
Description
- Molecular weight: 1,009.9 Da
- Purity: >95%
- Form: Solid
- Cell-permeable, ratiometric, fluorescent Ca2+ indicator
- High affinity binding to Ca2+ (Kd = 230 nM)
- Fluorescence increases upon Ca2+ binding (blue/violet emission)
- Emission and excitation shifts: 475 to 401 nm and 346 to 330 nm, depending on Ca2+ binding
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Substrate | Ca2+ |
| Target Molecule | Calcium |
| Shipping Condition | Ambient |
| Form | Solid |
| Detection Location | In-Cell Detection |
| Detection Method | Fluorescence |
| Content And Storage | Store at -20°C. |
| Buffer | 100% acetyloxymethyl 2-[4-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-3-[2-[2-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-5-methylphenoxy]ethoxy]phenyl]-1H-indole-6-carboxylate |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.