Learn More
Abcam Indo-1 K+ Salt, Fluorescent Ca2+ Indicator, 1 mg
Description
- Purity: >95%
- Molecular Weight: 840.1 Da
- CAS Number: 132319-56-3
- Properties: Cell-impermeable, fluorescent, ratiometric Ca2+ indicator
- Binding Affinity: Kd = 230 nM for Ca2+
- Excitation and Emission: Excitation shifts from 346 to 330 nm and emission shifts from 475 to 401 nm depending on Ca2+ binding
- Composition: 100% Indo-1 pentapotassium salt
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Calcium (Ca2+) |
| Substrate Properties | Chemical Substrate |
| Substrate | Ca2+ |
| Shipping Condition | Ambient |
| Buffer | 100% Indo-1 pentapotassium salt |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Detection Method | Fluorescence |
| Formulation | 100% Indo-1 pentapotassium salt |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.