Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indocyanine Green, Analytical Reagent, MP Biomedicals

Catalog No. ICN210667 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210667 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210667 Supplier MP Biomedicals, Inc Supplier No. 0521066750
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical reagent grade indocyanine green in powder form for laboratory research applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3599-32-4
Molecular Formula C43H47N2NaO6S2
Molecular Weight (g/mol) 774.967
MDL Number MFCD00013078
InChI Key MOFVSTNWEDAEEK-UHFFFAOYSA-M
Synonym Cardiogreen, Foxgreen, IC Green
PubChem CID 132274068
ChEBI CHEBI:31696
IUPAC Name sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate
SMILES CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+]

Specifications

Color Green
Quantity 50 mg
Physical Form Powder
Formula Weight 774.98
Grade Reagent
Chemical Name or Material Indocyanine Green
Recommended Storage Store at Room Temperature (15 to 30°C), desiccate.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.