accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CCH3NSOOOH3CH3CNSOOO(E)(E)(E/Z)(E/Z)

CAS RN 3599-32-4

IUPAC Name: sodium 2-[(5E)-7-[(2E)-1,1-dimethyl-3-(4-sulfonatobutyl)-1H,2H,3H-benzo[e]indol-2-ylidene]hepta-1,3,5-trien-1-yl]-1,1-dimethyl-3-(4-sulfonatobutyl)-1H-benzo[e]indol-3-ium
Synonyms: indocyanine green
Molecular Weight (g/mol): 774.97
Molecular Formula: C43H47N2NaO6S2
InChi Key: MOFVSTNWEDAEEK-UHFFFAOYSA-M
SMILES: [Na+].CC1(C)\C(=C/C=C/C=CC=CC2=[N+](CCCCS([O-])(=O)=O)C3=CC=C4C=CC=CC4=C3C2(C)C)N(CCCCS([O-])(=O)=O)C2=CC=C3C=CC=CC3=C12
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (1)
  • (3)
  • (1)
  • (4)
Color 
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (3)
  • (5)
  • (4)
  • (4)
  • (2)