Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indocyanine green, MP Biomedicals™

Catalog No. ICN15502050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15502050 50 mg
ICN15502005 5 mg
ICN15502025 25 mg
ICN15502080 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15502050 Supplier MP Biomedicals, Inc Supplier No. 0215502050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3599-32-4 | C43H47N2NaO6S2 | 774.967 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3599-32-4
Molecular Formula C43H47N2NaO6S2
Molecular Weight (g/mol) 774.967
MDL Number MFCD00013078
InChI Key MOFVSTNWEDAEEK-UHFFFAOYSA-M
Synonym Cardiogreen, Fax green
PubChem CID 132274068
ChEBI CHEBI:31696
IUPAC Name sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate
SMILES CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+]

Specifications

Assay Percent Range ∽ 90%
Color Green
pH 6
Quantity 50 mg
Physical Form Crystalline Powder
Formula Weight 774.98
Percent Purity ∼90%
Grade Research
Chemical Name or Material Indocyanine green
Recommended Storage Store at Room Temperature (15 to 30°C). Desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.