Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indoine Blue, MP Biomedicals

Catalog No. ICN202620 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202620 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202620 Supplier MP Biomedicals, Inc Supplier No. 0520262001

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4569-88-4
Molecular Formula C30H24ClN5O
Molecular Weight (g/mol) 506.01
MDL Number MFCD00011924
InChI Key SCMDRBZEIUMBBQ-UHFFFAOYSA-N
PubChem CID 9575841
IUPAC Name 3-amino-2,8-dimethyl-7-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride
SMILES [Cl-].CC1=CC2=NC3=CC(C)=C(N)C=C3[N+](C3=CC=CC=C3)=C2C=C1N\N=C1/C(=O)C=CC2=CC=CC=C12

Specifications

Quantity 1 g
Chemical Name or Material Indoine Blue
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.