Learn More
Abcam Indole-3-Butyric Acid Potassium Salt, Auxin-Family Plant Hormone, >98% Purity, Solid, 25 g
Description
- Molecular weight: 241.33 g/mol
- Purity: >98%
- Form: Solid
- Composition: 100% Indole-3-butyric acid potassium salt
- Description: Water soluble auxin-family plant hormone, considered a precursor to indole-3-acetic acid (IAA)
- CAS Number: 60096-23-3
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Indole-3-butyric Acid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Quantity | 25 g |
| For Use With (Application) | Plant hormone research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.