Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole-5-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. I067425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I067425G 25 g
I06745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I067425G Supplier TCI America Supplier No. I067425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1196-69-6
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD02093664
InChI Key ADZUEEUKBYCSEY-UHFFFAOYSA-N
Synonym indole-5-carboxaldehyde, 5-formylindole, indole-5-carbaldehyde, 1h-indole-5-carboxaldehyde, 5-indolealdehyde, indole-5-aldehyde, 5-formyl-1h-indole, 5-indolecarboxaldehyde, indole-5-carboxaldehyde 5-formylindole, 5-formyl-indole
PubChem CID 589040
IUPAC Name 1H-indole-5-carbaldehyde
SMILES C1=CC2=C(C=CN2)C=C1C=O

Specifications

Melting Point 100°C
Quantity 25 g
Formula Weight 145.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Indole-5-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.