Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole 99.0+%, TCI America™
SDP

Catalog No. I0021100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0021100G 100 g
I002125G 25 g
I0021500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0021100G Supplier TCI America Supplier No. I0021100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 120-72-9
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.151
MDL Number MFCD00005607
InChI Key SIKJAQJRHWYJAI-UHFFFAOYSA-N
Synonym indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german
PubChem CID 798
ChEBI CHEBI:16881
IUPAC Name 1H-indole
SMILES C1=CC=C2C(=C1)C=CN2

Specifications

Melting Point 53°C
Color White
Boiling Point 253°C
Quantity 100 g
UN Number 2811
Formula Weight 117.15
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Indole
EINECSNumber (5)-0130
RTECSNumber NL2450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.