Learn More
Selleck Chemical LLC Brivudine (BVDU) S5009-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S500910MM/1ML
Encompass_Preferred
Brivudine (BVDU) is a uridine derivative and nucleoside analog with pro-apoptotic and chemosensitizing properties It is incorporated into the viral DNA and blocks the action of DNA polymerases thus inhibiting viral replication
Catalog No. 50-311-0170
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.