Learn More
Medchemexpress LLC HY-14397A 500mg Medchemexpress, Indomethacin (sodium hydrate) CAS:74252-25-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14397A500MG
Encompass_Preferred
Medchemexpress, HY-14397A 500mg Indomethacin (sodium hydrate) CAS:74252-25-8 Indomethacin sodium hydrate (Indometacin sodium hydrate) is a potent, blood-brain permeable and nonselective inhibitor of COX1 and COX2, with IC50s of 18 nM and 26 nM for human COX-1 and COX-2, respectively, in CHO cells. Indomethacin sodium hydrate disrupts autophagic flux by disturbing the normal functioning of lysosomes[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0530
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.