Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indophenol Sodium Salt 95%, Thermo Scientific™

Catalog No. AC412340050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC412340050 5 g
AC412340250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC412340050 Supplier Thermo Scientific Chemicals Supplier No. 412340050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5418-32-6
Molecular Formula C12H8NNaO2
Molecular Weight (g/mol) 221.191
InChI Key FRUIYGAEJNTDCA-UHFFFAOYSA-M
Synonym indophenol sodium salt, indophenol, sodium salt, n-4-sodiooxyphenyl-p-benzoquinone monoimine, sodium 4-4-oxocyclohexa-2,5-dien-1-ylidene amino phenolate, 2,5-cyclohexadien-1-one, 4-4-hydroxyphenyl imino-, sodium salt, 4-p-hydroxyphenyl-2,5-cyclohexadiene-1-one sodium salt, 2,5-cyclohexadien-1-one, 4-p-hydroxyphenyl imino-, sodium salt, 2,5-cyclohexadien-1-one, 4-4-hydroxyphenyl imino-, sodium salt 1:1
PubChem CID 2735060
IUPAC Name sodium;4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
SMILES C1=CC(=O)C=CC1=NC2=CC=C(C=C2)[O-].[Na+]

Specifications

Quantity 5 g
Formula Weight 221.19
Chemical Name or Material Indophenol sodium salt, 95%
EINECSNumber 226-524-1
RTECSNumber GU5780000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.