Learn More
Cayman Chemical GSK121trIfluoROCTAte salt 10mg
SDP
Supplier:  Cayman Chemical 1749110 MG
Encompass_Preferred
A 13R14S-dihydroxy DHA formed by recombinant human macrophage 12-LO and sEH co-incubated with DHA reduces neutrophil infiltration in a mouse model of peritonitis and enhances human macrophage phagocytosis of zymosan A
Catalog No. 50-296-7753
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.