Learn More
Impacto™ Industrial Compression Energy Socks
Catalog No. 19400140 Shop All Impacto Protective Products Products
Encompass_Preferred
Change view
Click to view available options
Size:
Large; Men: 10.5 to 12
Medium; Men: 7.5 to 10
X to Large; Men: 12.5 to 14
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-400-140 X to Large; Men: 12.5 to 14
19-400-138 Large; Men: 10.5 to 12
19-400-139 Medium; Men: 7.5 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-400-140 Supplier Impacto™ Supplier No. NRGSOCKXL
Impacto™ NRGSOCK Industrial Compression Energy Socks provide support for long term standing on concrete floors. Ideal for extended shift operations: 10-12 hours.
Impacto™ Compression Industrial Socks
  • Offer moderate graduated compression 15-20mmHg for quicker blood return
  • Cushioned in Achilles tendon area for better protection and comfort
  • Breathable foot with arch support
  • Reciprocated heel and toe ensure better fit and comfort
  • Helpful with other medical conditions such as Deep Vein Thrombosis (DVT), varicose veins, circulation issues associated with diabetes and leg and ankle swelling
  • Sizing - Ankle Circumference L 10-11.5 in; XL 11.5-13 in. Calf Circumference L 16.5-18 in; XL 18-19.75
Specifications
Color Black
Material Nylon
Size X to Large; Men: 12.5 to 14
Toe Material Plain Toe
Gender Unisex
Product Type Socks