Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Inertia Demonstrator
SDPSCI_ED

Catalog No. S139494 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139494 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139494 Supplier United Scientific™ Supplier No. IDEM01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A classic demonstration for students to easily understand inertia.

This durable inertia demonstrator works reliably every time and is a great improvement on traditional versions of this classic experiment.

  • A steel ball rests on a thin metal card on top of a pillar, and a spring-loaded catch is mounted next to the pillar
  • When the catch is released, a piston strikes the edge of the metal card sharply
  • The card flies out from under the ball, which remains in place on top of the pillar due to its large inertial mass
  • The card is tethered to the aluminum base for easy recovery after the experiment
  • In a second experiment the cord is pulled slowly and steadily
  • In this case, the ball and card fall off the pillar together
  • Includes activity guide
  • Approximate product dimensions are 5 x 3 x 2 in.

Specifications

Grade 5 to 12
Format Kit
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.