Learn More
GL Sciences Inc Inertsil SIL-100A Analytical Column, 5um, 4.6mm ID, 250mm
Supplier:  GL Sciences Inc 502001712
Encompass_Preferred
Inertsil SIL-100A is a pure silica gel column available in normal phase mode. Because of the high quality of its silica gel, Inertsil SIL-100A achievs separation with sharp peaks and provides high column-to-column reproducibility. This excellent silica gel ideally designed for HPLC is the basis for “Inertsil 3-series” of GL Scineces. GL Sciences is the first company which emphasized the importance of silica-gel purity and determined the nature of the silanol impurities in the Silica gel.
Catalog No. NC1831774
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.