Learn More
Fisherbrand™ Infecon™ 3000 Infectious Substance Shipper
Catalog No. 19037437
Change view
Click to view available options
Type:
Bubble Pouch Absorbent Strips
Bubble Wrap and Absorbent Strips
Infectious Substance Shipper
Pressure Vessels
Refurbishment
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
19-037-437 Refurbishment
19-038-300B Bubble Pouch Absorbent Strips
19-038-300C Pressure Vessels
19-038-300A Bubble Wrap and Absorbent Strips
19-037-436 Infectious Substance Shipper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-037-437 Supplier Fisherbrand™ Supplier No. INF3001
Add to Cart
Add to Cart
Combines U.N. certification requirements with new innovative safety features
  • Each shipper consists of a transparent pressure vessel able to withstand up to 95kPa, cushioning materials, outer box, labels, declarations and instructions
  • Transparent pressure vessel allows contents to be seen
  • Use the Infecon 3000 with the Speci-Freez™ Insulated Shipper (19-075-384C) for refrigerated or frozen specimens
  • Reusable when using the complete refurbishment kit (19-037-437)
Order Info
Dimensions: 6 x 6 x 8 in. (15.24 x 15.24 x 20.32cm) with 1L vessel
Specifications
Applications For infectious substances
Type Refurbishment