Learn More
Com-Pac International™ Infecon Infectious Substance Shipper
Com-Pac International™ Infecon Infectious Substance Shipper has the best features of both an overpack shipper (INFECON 6000) and a UN Certified Category A shipper (INFECON 2000).
$329.00 - $482.50
Specifications
Quantity 4/case
Dimensions (L x W x H) Exterior 15.5 x 14 x 12.75 in.
View More Specs
Products 2
Catalog Number Mfr. No. Capacity Includes Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Includes Price Quantity & Availability  
22-043-316
View Documents
Com-Pac International™
INF6200
Holds up to 6 standard blood tubes Cooler, Shipping Box, Stabilizer Sheet, Instruction Sheets, Package Return Label, Dry Ice Super Label, Infectious Substance Dry Ice Super Label, 1/2 Liter Pressure Vessel, 2 x 2 Biological Substance Label, 2 x 2 Infectious Substance Label, Shippers Declarations, 50 mL Capacity Absorbent Sheet 3 x 3, Perforated 6 Compartment Bubble Pouch
Case of 4 for $329.00
Only null left
Add to Cart
22-043-317
View Documents
Com-Pac International™
INF6300
Holds up to 20 standard blood tubes Cooler, Shipping Box, Instruction Sheets, Package Return Label, Dry Ice Super Label, Infectious Substance Dry Ice Super Label, 1 Liter Pressure Vessel, 6 x 7 Mini Super Infectious Substance Label, Shippers Declarations, 50 mL Capacity Absorbent Sheet 3 x 3, Perforated 6 Compartment Bubble Pouch, Foam Insert
Case of 4 for $482.50
Only null left
Add to Cart
Description
Description
  • Combination of both units and provides a shipping solution for any Category A or B specimen.
  • Vessel in its carton is to be placed in the overpack cooler, with refrigerated gel packs or dry ice.
  • Designed to ship Ebola, Enterovirus, Zika Virus, and Measles.
  • Included in the kit are specific instructions for shipment of Ebola and Enterovirus.
  • Can, however, be used to ship any Category A or B specimen.
  • Temperature protection for up to 76 hours
  • Holds approximately 22 pounds of Dry Ice
  • Expanded Polystyrene Insulated Tub
  • Sturdy Outer Box
  • Infectious Substance
  • Dry Ice Super Label
Specifications
Specifications
4/case
15.5 x 14 x 12.75 in.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.