Learn More
Saf T Pak Inc STP-130 - Category Ambient Shipping System (UN 2900, UN 2814)
Supplier: Saf T Pak Inc STP130
Ambient Infectious Shipper, Inner box: 6.625L x 3.375W x 3.375H iniches, Outer box: 11L x 7.75W x 6.75H iniches, Components: Reusable 4G outer box, Inner box, Instruction manual, Hazard labels, 10/Case
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.