Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Infectious pancreatic necrosis virus ELISA Kit

Catalog No. 89032340 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-340 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-340 Supplier Abnova™ Supplier No. KA4892
Add to Cart
Add to Cart

Enzyme-linked immunosorbent assay for the detection of infectious pancreatic necrosis virus (IPNV)

  • Used for the in vitro quantitative determination of human SOD1 in human serum, human plasma, cell lysate, culture supernatants and buffered solution
  • Assay will recognize native and recombinant human IL-9

Specifications

Description Infectious pancreatic necrosis virus ELISA Kit is an enzyme-linked immunosorbent assay for the detection of infectious pancreatic necrosis virus (IPNV).
Conjugate HRP
Detection Method Colorimetric
Product Type Infectious pancreatic necrosis virus ELISA Kit
Sample Type Cell Culture Media, Homogenate
Target Species Bovine
For Use With (Application) Qualitative
Kit Contents Microtitre Plate: 96 wells, Negative Antigen: 0.7mL, Positive Antigen: 0.7mL, RAIPNV: 6mL, Conjugate: 12mL, TMB-Complete 5: 15mL, Wash Solution: 20x concentrated buffer (20mL), Stop Solution: Acid solution ready to use (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 10 μL (Prediluted)
Storage Requirements Store the kit at 4°C. Do not freeze.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.