Learn More
Enzo Life Sciences Infigratinib (free base) (25mg). CAS: 872511-34-7
SDP
Supplier:  Enzo Life Sciences ENZCHM2250025
Encompass_Preferred
Infigratinib is a pan FGFR kinase inhibitor, and is an orally bioavailable pan inhibitor of human fibroblast growth factor receptors (FGFRs) with potential antiangiogenic and antineoplastic activities. Alternative name: BGJ-398. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3605
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.