Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the estimation of Infliximab (REMICADE) in serum and plasma. Calibrated against international WHO standard 16/170 from NIBSC. Assay Range: 10 - 160 ng/ml; Sensitivity: 8 ng/ml.
Supplier:  KRISHGEN BIOSYSTEMS KBI1011
Encompass_Preferred
Pack Size: 1 x 96 wells. This mAb-based Infliximab ELISA is optimised to detect infliximab and biosimilars in human samples. If you would like to use animal samples, please contact us. The Infliximab ELISA is validated against ICH M10 guidelines for bioassays. Background: Infliximab is a monoclonal antibody which has high specificity for TNF alpha, and does not neutralize TNF beta. Infliximab causes programmed cell death of TNF alpha expressing activated T lymphocytes, an important cell type mediating inflammation; hence it is generally assumed that resolution of activated T cells by Infliximab explains its efficacy in Crohns disease. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Precision CV <10%.
Catalog No. NC2747147
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.