Learn More
MyBioSource Human Interleukin 1 beta (IL1B) ELISA Kit, 96 Tests
SDP
Supplier:  MyBioSource MBS59006496STRIPWELLS
Encompass_Preferred
The Human IL-6 ELISA Kit quantifies IL-6 in various biological fluids using a 96-well plate coated with an antibody specific to IL-6. Samples are added, followed by a biotinylated detection antibody, HRP-conjugated streptavidin, and TMB substrate for color development, which is measured at 450 nm.
Catalog No. NC2050826
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.