Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A H1N1 HA (A/Puerto Rico/8/34) Recombinant Rabbit Monoclonal Antibody (016)

Catalog No. MA560327
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA560327 20 μL
MA560326 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA560327 Supplier Invitrogen™ Supplier No. MA560327
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recognizes Influenza H1N1 (A/Puerto Rico/8/1934) Hemagglutinin/HA; No cross-reactivity in ELISA with: H1N1 (A/Victoria/4897/2022) HA Protein; H1N1 (A/Wisconsin/588/2019)/(A/Victoria/2570/2019) HA Protein; H1N1 (A/Wisconsin/67/2022) HA Protein; H3N2 (A/Darwin/6/2021) HA Protein; H3N2 (A/Darwin/9/2021) HA Protein; Influenza B (B/Austria/1359417/2021) HA Protein; Influenza B (B/PHUKET/3073/2013) HA Protein; Influenza B (B/Washington/02/2019) HA Protein.

Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus was identified in specimens obtained from patients in Mexico and the United States. The virus spread quickly around the world and on June 11, 2009, the World Health Organization declared it a pandemic. Influenza A virus has one of sixteen possible Hemagglutinin (HA) surface proteins and one of nine possible Neuraminidase (NA) surface proteins. The Hemagglutinin protein facilitates viral attachment while Neuraminidase is involved in viral release. These proteins also elicit immune responses that prevent infection or independently reduce viral replication. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin. The distinct antigenic properties of the new swine influenza virus compared with seasonal influenza A (H1N1) virus suggest that human immunity against new swine influenza virus is limited, although the age distribution of reported cases suggests some degree of protection in older age groups.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H1N1 HA (A/Puerto Rico/8/34)
Applications ELISA
Classification Recombinant Monoclonal
Clone 016
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias flu hemagglutinin; H1N1 HA; H1N1 hemagglutinin; Influenza H1N1; influenza hemagglutinin
Host Species Rabbit
Immunogen Recombinant Influenza H1N1 (A/Puerto Rico/8/1934) Hemagglutinin/HA protein (Met1-Gln528).
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.