Learn More
Sino Biological INFLUENZA A H3N2 A DARWIN 6 2
Supplier:  Sino Biological 41051V27HB
Encompass_Preferred
Influenza A H3N2 (A/Darwin/6/2021) Hemagglutinin / HA-specific B cell probe Protein (Y114F, AVI & His Tag), Biotinylated (HPLC-verified)
Catalog No. NC3654261
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.