Learn More
Description
Specifications
Specifications
| Assay Range | 390.63 pg/mL |
| Assay Sensitivity | 390.63-25000 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Influenza A H3N2 (A/Brisbane/10/2007). |
| Target Species | Influenza A Virus H3N2 |
| For Use With (Application) | Sandwich ELISA |
| Host Species | Mouse/Rabbit |
| Kit Contents | |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
