Learn More
Influenza A H5N8 Hemagglutinin Antibody (HL1550) - Azide and BSA Free, R&D Systems™

Description
Specifications
Specifications
| Antigen | Influenza A H5N8 Hemagglutinin |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | HL1550 |
| Conjugate | Unconjugated |
| Dilution | Western Blot -Assay dependent, Immunohistochemistry -Assay dependent, Immunohistochemistry-Paraffin Assay dependent |
| Formulation | PBS |
| Gene Alias | Avian Influenza A H5N8 Hemagglutinin, Haemagglutinin, influenzavirus A, Influenza A H5N8 HA, Influenza A Haemagglutinin, Influenza A Hemagglutinin |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of Influenza A virus H5N8 HA (Hemagglutinin)(A/Astrakhan/3212/2020)). The exact sequence is proprietary. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.