Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A H6N1 NP Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675733 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-733 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-733 Supplier Invitrogen™ Supplier No. PA141156
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Influenza A H6N1 NP Polyclonal Antibody for ICC/IF, ELISA

An H6N1 influenza virus was isolated from a green-winged teal during the H5N1 outbreak in Hong Kong Special Administrative Region (SAR) in 1997. This virus possesses similar genes encoding internal proteins as in the human H5N1 and H9N2 influenza viruses. In 1999, influenza viruses from quail infected two humans in Hong Kong. suggesting the potential for avian influenza viruses to cross the species barrier and infect humans without prior reassortment in an intermediate host, such as the pig. The common features shared by H5N1 and H9N2 influenza viruses isolated from humans are the genes encoding the proteins of the replicating complex, the matrix protein (M) gene, the nonstructural protein (NS) gene, N1 neuraminidase (NA), This virus essentially represents the reemergence of the H5N1 influenza viruses with a different hemagglutinin (HA).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H6N1 NP
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Alias H6N1 NP; Influenza H6N1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 428-441 of avian flu nucleoprotein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.