Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H7N7 Hemagglutinin Rabbit anti-Virus, HRP, Antibody Pair, Novus Biologicals™

Antibody pair for Solid phase sandwich ELISA
$458.00 - $1124.00
Specifications
Antigen | Influenza A H7N7 Hemagglutinin |
---|---|
Applications | ELISA |
Conjugate | HRP |
Host Species | Rabbit |
Regulatory Status | RUO |
Description
Solid phase sandwich ELISA for quantitative determination of Influenza A H7N7 HemagglutininSpecifications
Influenza A H7N7 Hemagglutinin | |
HRP | |
RUO | |
Storage of components varies. See protocol for specific instructions. |
ELISA | |
Rabbit | |
Virus | |
Antibody Pairs |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title