Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A H7N9 HA Recombinant Rabbit Monoclonal Antibody (001)

Catalog No. MA560313
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA560313 20 μL
MA560312 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA560313 Supplier Invitrogen™ Supplier No. MA560313
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Has cross-reactivity in ELISA with: H7N9 (A/Anhui/1/2013) HA Protein; H7N9 (A/Hangzhou/1/2013) HA Protein; H7N9 (A/Hangzhou/3/2013) HA Protein; H7N9 (A/Pigeon/Shanghai/S1069/2013) HA Protein; H7N9 (A/Shanghai/1/2013) HA Protein; H7N9 (A/Shanghai/2/2013) HA Protein; H7N9 (A/Shanghai/4664T/2013) HA Protein; H7N9 (A/Zhejiang/1/2013) HA Protein; H7N9 (A/Zhejiang/DTID-ZJU10/2013) HA Protein; H7N7 (A/chicken/Netherlands/1/03) HA Protein; H7N7 (A/Netherlands/219/2003) HA Protein; H7N2 (A/ruddy turnstone/New Jersey/563/2006) HA Protein; H7N3 (A/turkey/Italy/214845/2002) HA Protein; H7N8 (A/mallard/Netherlands/33/2006) HA Protein; H10N3 (A/duck/Hong Kong/786/1979) HA Protein; H15N8 (A/duck/AUS/341/1983) HA Protein; No cross-reactivity in ELISA with: H1N1 (A/California/07/2009) HA; H1N1 (A/California/04/2009) HA0 Protein; H2N2 (A/Canada/720/2005) HA Protein; H3N2 (A/Brisbane/10/2007) HA Protein; H4N6 (A/mallard/Ohio/657/2002) HA Protein; H5N1 (A/Anhui/1/2005) HA Protein; H6N1 (A/northern shoveler/California/HKWF115/2007) HA Protein; H7N1 (A/turkey/Italy/4602/99) HA1 Protein; H7N3 (A/chicken/SK/HR-00011/2007) HA1 Protein; H7N7 (A/equine/Kentucky/1a/1975) HA Protein; H7N7 (A/Netherlands/219/2003) HA Protein; H7N7 (A/Netherlands/219/2003) HA1 Protein; H8N4 (A/pintail duck/Alberta/114/1979) HA Protein; H9N2 (A/Hong Kong/1073/99) HA Protein; H11N2 (A/duck/Yangzhou/906/2002) HA Protein; H12N5 (A/green-winged teal/ALB/199/1991) HA Protein; H13N8 (A/black-headed gull/Netherlands/1/00) HA Pr...

Influenza A H7N9 HA refers to the hemagglutinin (HA) protein found on the surface of the Influenza A H7N9 virus. Hemagglutinin is one of the major glycoproteins on the influenza virus surface and plays a crucial role in the virus's ability to infect host cells. It facilitates the binding of the virus to the host cell receptors and subsequent entry into the cell. The 'H7N9' designation indicates the specific subtype of the influenza virus, where 'H7' refers to the type of hemagglutinin and 'N9' refers to the type of neuraminidase, another surface protein involved in the release of new virus particles from infected cells. Influenza A H7N9 first emerged in humans in China in 2013 and has been associated with severe respiratory illness and high mortality rates. The HA protein is a primary target for the immune response and is critical for the development of vaccines and antiviral drugs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H7N9 HA
Applications ELISA
Classification Recombinant Monoclonal
Clone 001
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias flu hemagglutinin; influenza hemagglutinin
Host Species Rabbit
Immunogen Recombinant Influenza A H7N9 HA protein.
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.