Learn More
Influenza A H9N2 Hemagglutinin anti-Virus, HRP, Antibody Pair, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Influenza A H9N2 Hemagglutinin |
| Applications | ELISA, Sandwich ELISA |
| Conjugate | HRP |
| Gene Alias | HA, Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain |
| Quantity | 15 Plates |
| Regulatory Status | RUO |
| Target Species | Virus |
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Product Type | Antibody Pairs |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.