Learn More
Novus Biologicals™ Influenza A Haemagglutinin H1N1 Antibody Pair [HRP]
Shop All R&D Systems Products

Description
Specifications
Specifications
| Assay Range | 46.88 pg/mL |
| Assay Sensitivity | 46.88-3000 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Influenza A H1N1 (A/Puerto Rico/8/1934) Haemagglutinin. |
| Target Species | Virus |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | HA, HA1, HA2, Hemagglutinin, Influenza A Virus Hemagglutinin |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.