Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A M1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532253
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532253 Supplier Invitrogen™ Supplier No. PA532253
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: DF-1, WSN virus infected DF-1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Influenza A virus matrix 1, M1, is a critical protein required for assembly and budding. Hemagglutinin associates with M1 via its cytoplasmic tail and transmembrane domain. The M2 and nucleoprotein proteins are critical in the replication cycle of influenza viruses. The M2 channel protein is an essential component of the viral envelope because of its ability to form a highly selective, pH-regulated, proton-conducting channel. The M2 channel allows protons to enter the interior of the virus and acidification weakens the interaction of the M1 protein with the ribonuclear core.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A M1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene Alias Influenza M1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Internal region of Influenza A virus M1 (matrix protein) (A/Puerto Rico/8/1934(H1N1)). The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.