Learn More
Invitrogen™ Influenza A NP Monoclonal Antibody (2F205)

Description
Influenza A NP Monoclonal Antibody for ELISA
Specifications
Specifications
| Antigen | Influenza A NP |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2F205 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene Alias | Influenza nucleoprotein |
| Host Species | Mouse |
| Immunogen | Recombinant influenza A H1N1 protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.