Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A NP Monoclonal Antibody (H72G)
GREENER_CHOICE

Catalog No. MA17323
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17323 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17323 Supplier Invitrogen™ Supplier No. MA17323
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7323 antibody reacts with Influenza A virus NP in viral and infected tissue samples. MA1-7323 has been successfully used in Immunofluorescence and ELISA applications. The MA1-7323 antibody was raised against Influenza A Nucleoprotein.

The nucleoprotein (NP) of Influenza A virus encapsidates the negative strand of the viral RNA and is essential for replicative transcription. It may also be involved in other essential functions throughout the virus life cycle. As well as binding ssRNA, NP is able to self associate to form large oligomeric complexes. NP is able to interact with a variety of other macromolecules of both viral and cellular origins. It binds the PB1 and PB2 subunits of the polymerase and the matrix protein M1. NP has also been shown to interact with at least four cellular polypeptide families: nuclear import receptors of the importin class, filamentous (F) actin, the nuclear export receptor CRM1 and a DEAD box helicase BAT1/UAP56 (Portela et al 2002).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A NP
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone H72G
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Influenza nucleoprotein
Host Species Mouse
Immunogen Influenza A Nucleoprotein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.