Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A PB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532220 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532220 Supplier Invitrogen™ Supplier No. PA532220
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: Influenza virus (A/WSN/1933(H1N1))-infected DF-1 cell lysates. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Influenza A virus contains eight single stranded, negative sense RNAs. Each RNA is associated with viral RNA-dependent RNA polymerase which consists of PA, PB1 and PB2 proteins. PB1 is the subunit involved in the catalytic activity of nucleotide polymerization and is involved in the initiation of transcription. Both PB1 and PB2 can be crosslinked to synthetic RNA with the 3' terminal sequence of viral RNA. PB1 and PB2 may also be involved in recognition of the promoter and/or replication origin on template viral RNA. PB2 is the cap 1-recognition protein, which binds to the cap structure of host mRNA. The cap structure then acts as a primer for transcription to produce viral mRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A PB2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Alias Influenza PB2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of Influenza A virus PB2 (A/Puerto Rico/8/1934(H1N1)). The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.