Learn More
Creative Biomart PEV3A
Supplier:  Creative Biomart VAC0375
Encompass_Preferred
PEV3A is a prophylactic malaria vaccine that acts against various stages of the disease. It is a two-component vaccine that contains the virosome formulated peptide mimetics PEV301 and PEV302. The two
Catalog No. 50-208-5215
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.